ENA053247283 Enamine

2'-cyano-[1,1'-biphenyl]-2-carboxylic acid

  • MDL number MFCD00512056
  • Cas Number : 57743-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-174
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13212
Availability details for
Quantity
  • Availability
  • Add to Cart