ENA053247293 Enamine

(9-chloro-3,4-dihydro-2H-1,5-benzodioxepin-7-yl)methanol

  • MDL number MFCD06655730
  • Cas Number : 852956-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-46
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13222

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13222
Availability details for
Quantity
  • Availability
  • Add to Cart