ENA053247295 Enamine

1-amino-4,6-dimethyl-5-(methylsulfanyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD06655731
  • Cas Number : 120456-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13224
Availability details for
Quantity
  • Availability
  • Add to Cart