ENA053247331 Enamine

N-[2-(allylthio)phenyl]-2-chloroacetamide

  • MDL number MFCD06655753
  • Cas Number : 79877-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13262

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13262
Availability details for
Quantity
  • Availability
  • Add to Cart