ENA053247333 Enamine

1-(methylsulfonyl)-1,2,3,4-tetrahydroquinoline-6-carboxylic acid

  • MDL number MFCD06655754
  • Cas Number : 852916-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218...220
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13264

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13264
Availability details for
Quantity
  • Availability
  • Add to Cart