ENA053247342 Enamine

N-[4-(aminosulfonyl)benzyl]acetamide

  • MDL number MFCD00547183
  • Cas Number : 2015-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13273

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13273
Availability details for
Quantity
  • Availability
  • Add to Cart