ENA053247348 Enamine

N-cyclopropyl-2-(methylamino)acetamide

  • MDL number MFCD06655764
  • Cas Number : 852916-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13279
Availability details for
Quantity
  • Availability
  • Add to Cart