ENA053247350 Enamine

4-(2,5-dimethylthiophen-3-yl)-1,3-thiazol-2-amine

  • MDL number MFCD03424581
  • Cas Number : 436152-83-9
  • Molecular Weight : 210.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13281

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13281
Availability details for
Quantity
  • Availability
  • Add to Cart