ENA053247351 Enamine

1-(5,6,7,8-tetrahydronaphthalen-2-yl)ethanamine

  • MDL number MFCD06655766
  • Cas Number : 91562-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13282

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13282
Availability details for
Quantity
  • Availability
  • Add to Cart