ENA053247355 Enamine

N'-(2-chloropropanoyl)benzohydrazide

  • MDL number MFCD06655769
  • Cas Number : 851115-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13286

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13286
Availability details for
Quantity
  • Availability
  • Add to Cart