ENA053247362 Enamine

2-chloro-N-{[(3-chloro-2-methylphenyl)carbamoyl]methyl}acetamide

  • MDL number MFCD06655772
  • Cas Number : 851116-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13293
Availability details for
Quantity
  • Availability
  • Add to Cart