ENA053247364 Enamine

4-[(3,5-dimethyl-4-nitro-1H-pyrazol-1-yl)methyl]benzoic acid

  • MDL number MFCD00852420
  • Cas Number : 400752-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13295

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13295
Availability details for
Quantity
  • Availability
  • Add to Cart