ENA053247366 Enamine

4-(1,3-benzothiazol-2-yl)-1,3-thiazol-2-amine

  • MDL number MFCD06655775
  • Cas Number : 54469-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 247-248
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13297

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13297
Availability details for
Quantity
  • Availability
  • Add to Cart