ENA053247378 Enamine

3-{2,5,7-trimethylpyrazolo[1,5-a]pyrimidin-6-yl}propanoic acid

  • MDL number MFCD06655785
  • Cas Number : 851116-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13309
Availability details for
Quantity
  • Availability
  • Add to Cart