ENA053247382 Enamine

2-[5-(thiophen-2-yl)-2H-1,2,3,4-tetrazol-2-yl]acetic acid

  • MDL number MFCD03052414
  • Cas Number : 36855-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-221
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12948
Availability details for
Quantity
  • Availability
  • Add to Cart