ENA053247383 Enamine

N-(3-tert-butyl-1,2-oxazol-5-yl)-2-chloroacetamide

  • MDL number MFCD06655789
  • Cas Number : 668980-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13315
Availability details for
Quantity
  • Availability
  • Add to Cart