ENA053247397 Enamine

7-(4-chlorophenyl)-5,6-dimethyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine

  • MDL number MFCD06655797
  • Cas Number : 72578-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13329

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13329
Availability details for
Quantity
  • Availability
  • Add to Cart