ENA053247408 Enamine

N-[4-(2-amino-1,3-thiazol-4-yl)phenyl]benzamide

  • MDL number MFCD01119367
  • Cas Number : 299441-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-223
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13340
Availability details for
Quantity
  • Availability
  • Add to Cart