ENA053247410 Enamine

1-methanesulfonyl-2-methyl-2,3-dihydro-1H-indole-5-carboxylic acid

  • MDL number MFCD06655804
  • Cas Number : 851116-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-151
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13342
Availability details for
Quantity
  • Availability
  • Add to Cart