ENA053247428 Enamine

4-(1,1-dimethylpropyl)cyclohexanamine

  • MDL number MFCD03422590
  • Cas Number : 208941-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 223...226
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13360

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13360
Availability details for
Quantity
  • Availability
  • Add to Cart