ENA053247446 Enamine

2-(chloromethyl)-1-methoxy-4-methylbenzene

  • MDL number MFCD06655823
  • Cas Number : 7048-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 9-10
Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13378

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
hcodes C
MSDS https://www.enaminestore.com/catalog/EN300-13378
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart