ENA053247447 Enamine

6-(chloromethyl)phenanthridine

  • MDL number MFCD06655824
  • Cas Number : 40484-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-134
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13379
Availability details for
Quantity
  • Availability
  • Add to Cart