ENA053247453 Enamine

3,4-dimethyl-5-[(4-methylpiperidin-1-yl)sulfonyl]benzoic acid

  • MDL number MFCD01417121
  • Cas Number : 494838-94-7
  • Molecular Weight : 311.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13385
Availability details for
Quantity
  • Availability
  • Add to Cart