ENA053247456 Enamine

3-chlorothieno[2,3-b]thiophene-2-carboxylic acid

  • MDL number MFCD01927113
  • Cas Number : 39244-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 241-242
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13389
Availability details for
Quantity
  • Availability
  • Add to Cart