ENA053247458 Enamine

4-fluoro-3-methyl-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD06655826
  • Cas Number : 852940-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13391

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13391
Availability details for
Quantity
  • Availability
  • Add to Cart