ENA053247463 Enamine

methyl 6-(carbamothioylamino)hexanoate

  • MDL number MFCD06655829
  • Cas Number : 98998-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13396
Availability details for
Quantity
  • Availability
  • Add to Cart