ENA053247464 Enamine

5-morpholin-4-yl-4-phenyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06655830
  • Cas Number : 94971-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13397

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13397
Availability details for
Quantity
  • Availability
  • Add to Cart