ENA053247468 Enamine

1-(3-ethoxypropyl)-2-sulfanyl-4,5-dihydro-1H-imidazol-5-one

  • MDL number MFCD06655834
  • Cas Number : 852940-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13401
Availability details for
Quantity
  • Availability
  • Add to Cart