ENA053247469 Enamine

2-mercapto-3-(tetrahydrofuran-2-ylmethyl)-3,5-dihydro-4H-imidazol-4-one

  • MDL number MFCD03422804
  • Cas Number : 832737-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13402

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13402
Availability details for
Quantity
  • Availability
  • Add to Cart