ENA053247473 Enamine

3-(7-chloro-4-oxo-2-sulfanyl-3,4-dihydroquinazolin-3-yl)propanoic acid

  • MDL number MFCD06655838
  • Cas Number : 852940-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-13406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13406
Availability details for
Quantity
  • Availability
  • Add to Cart