ENA053247484 Enamine

4-(chloromethyl)-1-(difluoromethoxy)-2-methoxybenzene

  • MDL number MFCD06655582
  • Cas Number : 852706-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12949
Availability details for
Quantity
  • Availability
  • Add to Cart