ENA053247494 Enamine

N-(4-hydroxyphenyl)-N,4-dimethylbenzamide

  • MDL number MFCD06655850
  • Cas Number : 1148-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13427

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13427
Availability details for
Quantity
  • Availability
  • Add to Cart