ENA053247497 Enamine

1-(4-amino-2-methylquinolin-3-yl)ethanone

  • MDL number MFCD04220752
  • Cas Number : 71993-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152...154
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13430

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13430
Availability details for
Quantity
  • Availability
  • Add to Cart