ENA053247519 Enamine

(7-chloro-1,3-benzodioxol-5-yl)methanol

  • MDL number MFCD06655862
  • Cas Number : 179110-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102...104
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13452

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13452
Availability details for
Quantity
  • Availability
  • Add to Cart