ENA053247526 Enamine

1-cycloheptyl-2-sulfanyl-4,5-dihydro-1H-imidazol-5-one

  • MDL number MFCD06655868
  • Cas Number : 853723-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-73
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13460

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13460
Availability details for
Quantity
  • Availability
  • Add to Cart