ENA053247527 Enamine

3-benzyl-2-mercapto-3,5-dihydro-4H-imidazol-4-one

  • MDL number MFCD00125264
  • Cas Number : 39123-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13461

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13461
Availability details for
Quantity
  • Availability
  • Add to Cart