ENA053247529 Enamine

3-(4-isobutyl-5-mercapto-4H-1,2,4-triazol-3-yl)propanamide

  • MDL number MFCD06655871
  • Cas Number : 853723-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13463

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13463
Availability details for
Quantity
  • Availability
  • Add to Cart