ENA053247530 Enamine

2-cyclopropaneamido-4,5-dimethylthiophene-3-carboxylic acid

  • MDL number MFCD06655583
  • Cas Number : 852933-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-151
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12951
Availability details for
Quantity
  • Availability
  • Add to Cart