ENA053247532 Enamine

4-(1,3-benzothiazol-2-yl)benzene-1,2-diamine

  • MDL number MFCD06655874
  • Cas Number : 132064-28-9
  • Molecular Weight : 241.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167-168
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13466
Availability details for
Quantity
  • Availability
  • Add to Cart