ENA053247536 Enamine

N-(2-fluorobenzyl)cyclopropanamine

  • MDL number MFCD04035532
  • Cas Number : 625435-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13470

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-13470
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart