ENA053247541 Enamine

6-amino-5-(chloroacetyl)-1-ethylpyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD06655881
  • Cas Number : 67130-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13475

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13475
Availability details for
Quantity
  • Availability
  • Add to Cart