ENA053247544 Enamine

2-[5-(2-chloroacetyl)thiophen-2-yl]acetamide

  • MDL number MFCD06655883
  • Cas Number : 853724-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-116
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-13478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13478
Availability details for
Quantity
  • Availability
  • Add to Cart