ENA053247546 Enamine

2,3-dihydro-1H-indene-5-carbohydrazide

  • MDL number MFCD06655885
  • Cas Number : 321195-95-3
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-165
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13480
Availability details for
Quantity
  • Availability
  • Add to Cart