ENA061488095 Enamine

1-(2-methylphenyl)-3-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD06655925
  • Cas Number : 956437-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13556
Availability details for
Quantity
  • Availability
  • Add to Cart