ENA061488096 Enamine

2-({[1-(1,3-dioxaindan-5-yl)ethylidene]amino}oxy)acetic acid

  • Cas Number : 855716-75-5
  • Molecular Weight : 237.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13573
Availability details for
Quantity
  • Availability
  • Add to Cart