ENA061488106 Enamine

1-chloro-4-[(2-isothiocyanatoethyl)sulfanyl]benzene

  • MDL number MFCD07366301
  • Cas Number : 38752-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13662

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13662
Availability details for
Quantity
  • Availability
  • Add to Cart