ENA061488107 Enamine

3-[(carbamoylmethyl)sulfamoyl]benzoic acid

  • MDL number MFCD07323902
  • Cas Number : 848290-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13698
Availability details for
Quantity
  • Availability
  • Add to Cart