ENA061488109 Enamine

2-[2-(2,4-dimethylphenyl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD07324810
  • Cas Number : 847744-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13724
Availability details for
Quantity
  • Availability
  • Add to Cart