ENA061488114 Enamine

4-({[(propan-2-yl)carbamoyl]amino}methyl)benzoic acid

  • MDL number MFCD07323326
  • Cas Number : 847744-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13751
Availability details for
Quantity
  • Availability
  • Add to Cart