ENA061488119 Enamine

3-(4,6-dimethyl-2-sulfanylpyrimidin-5-yl)propanoic acid

  • MDL number MFCD07324812
  • Cas Number : 847744-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13763
Availability details for
Quantity
  • Availability
  • Add to Cart